Learn More
Selleck Chemical LLC Benzoyleneurea S5166-25mg
SDP
Catalog No. 503112803
Encompass_Preferred
Benzoyleneurea (2 4-Dihydroxyquinazoline Quinazolinedione Quinazoline-2 4-diol) scaffold is used in the synthesis of novel protein geranylgeranyltransferase-I inhibitors Benzoyleneurea possesses anti-bacterial activity
Catalog No. 50-311-2803 Supplier Selleck Chemical LLC Supplier No. S516625MG
Benzoyleneurea (2 4-Dihydroxyquinazoline Quinazolinedione Quinazoline-2 4-diol) scaffold is used in the synthesis of novel protein geranylgeranyltransferase-I inhibitors Benzoyleneurea possesses anti-bacterial activity