Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoyleneurea | 86-96-4 | 1 G
SDP

Catalog No. 5000419666
Encompass_Preferred

Benzoyleneurea is a chemical compound with anti-bacterial activity. Its scaffold can be utilized in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors. This product appears as a white to yellow solid.

  • Possesses anti-bacterial activity
  • Scaffold for novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors synthesis
  • Molecular weight: 162.15
  • Chemical formula: C8H6N2O2
  • Appearance: Solid
  • Color: White to yellow
  • Purity: Typically 99.6% or higher

Catalog No. 50-004-19666 Supplier Medchemexpress LLC Supplier No. HYN70891G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzoyleneurea is a chemical compound with anti-bacterial activity. Its scaffold can be utilized in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors. This product appears as a white to yellow solid.

  • Possesses anti-bacterial activity
  • Scaffold for novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors synthesis
  • Molecular weight: 162.15
  • Chemical formula: C8H6N2O2
  • Appearance: Solid
  • Color: White to yellow
  • Purity: Typically 99.6% or higher

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.