Learn More
Medchemexpress LLC Benzoyleneurea | 86-96-4 | 1 ML
SDP
Catalog No. 5000420105
Encompass_Preferred
Benzoyleneurea is a chemical compound also known as quinazolinedione. It demonstrates antibacterial activity and its scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors. It is typically supplied as a white to yellow solid.
  • Exhibits antibacterial activity.
  • Scaffold useful for synthesizing novel PGGTase-I inhibitors.
  • Appears as a white to yellow solid.
  • High purity.
  • Soluble in DMSO.
  • Shipped at room temperature (continental US).
  • For research use only.
Catalog No. 50-004-20105 Supplier Medchemexpress LLC Supplier No. HYN708910MM/1ML
Benzoyleneurea is a chemical compound also known as quinazolinedione. It demonstrates antibacterial activity and its scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors. It is typically supplied as a white to yellow solid.
  • Exhibits antibacterial activity.
  • Scaffold useful for synthesizing novel PGGTase-I inhibitors.
  • Appears as a white to yellow solid.
  • High purity.
  • Soluble in DMSO.
  • Shipped at room temperature (continental US).
  • For research use only.