Learn More
Medchemexpress LLC Benzoyleneurea | 86-96-4 | 500 MG
SDP
Catalog No. 5000419687
Encompass_Preferred
Benzoyleneurea possesses anti-bacterial activity. The Benzoyleneurea scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors.
  • Possesses anti-bacterial activity.
  • Scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors.
Catalog No. 50-004-19687 Supplier Medchemexpress LLC Supplier No. HYN7089500MG
Benzoyleneurea possesses anti-bacterial activity. The Benzoyleneurea scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors.
  • Possesses anti-bacterial activity.
  • Scaffold can be used in the synthesis of novel protein geranylgeranyltransferase-I (PGGTase-I) inhibitors.