Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylferrocene, 98%, Thermo Scientific Chemicals

Catalog No. AAA1768503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1768503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1768503 Supplier Thermo Scientific Chemicals Supplier No. A1768503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1272-44-2
Molecular Formula C17H14FeO
Molecular Weight (g/mol) 290.14
MDL Number MFCD00001431
InChI Key JJHHJZCYDPLHIL-UHFFFAOYNA-N
Synonym Ferrocenyl phenyl ketone
IUPAC Name Benzoylferrocene
SMILES [Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1

Specifications

Melting Point 106°C to 109°C
Quantity 1 g
Formula Weight 290.14
Percent Purity 98%
Chemical Name or Material Benzoylferrocene
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-054-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.