Learn More
Cayman Chemical BenzoylhypaconIn 10mg
SDP
Catalog No. 502949015
Encompass_Preferred
A monoester diterpene alkaloid with anti-inflammatory activity; inhibits COX-2 activity in a cell-free assay (IC50 = 20.5 µM); reduces histamine-induced vascular permeability in rats at 200 mg/kg
Catalog No. 50-294-9015 Supplier Cayman Chemical Supplier No. 3404610 MG
A monoester diterpene alkaloid with anti-inflammatory activity; inhibits COX-2 activity in a cell-free assay (IC50 = 20.5 µM); reduces histamine-induced vascular permeability in rats at 200 mg/kg