Learn More
Cayman Chemical Benzoylmesaconine, Purity: ≥98%, CAS: 63238-67-5, 10 mg
SDP
Catalog No. 502949003
Encompass_Preferred
An alkaloid; promotes isolated rat liver mitochondrial energy metabolism (EC50 = 30.95 µg/ml); increases survival in a mouse model of burn-associated HSV1 infection at 1 µg/kg; increases the latency to tail withdrawal or a struggling response in a tail pressure test in mice (ED50 = 38.9 mg/kg)
Catalog No. 50-294-9003 Supplier Cayman Chemical Supplier No. 3323810 MG
An alkaloid; promotes isolated rat liver mitochondrial energy metabolism (EC50 = 30.95 µg/ml); increases survival in a mouse model of burn-associated HSV1 infection at 1 µg/kg; increases the latency to tail withdrawal or a struggling response in a tail pressure test in mice (ED50 = 38.9 mg/kg)