Learn More
Medchemexpress LLC Benzoylpaeoniflorin | 38642-49-8 | 584.57 | 100 MG
SDP
Catalog No. 5000416168
Encompass_Preferred
Benzoylpaeoniflorin is an orally active monoterpene glycoside compound that exerts anti-inflammatory, anti-allergic, psoriasis-improving, and sepsis-improving effects by inhibiting signaling pathways such as TNF/NF-κB and MAPK, as well as regulating immune homeostasis. It can be used in research related to immune, allergic, and inflammatory diseases.
  • Orally active monoterpene glycoside compound
  • Exerts anti-inflammatory, anti-allergic, psoriasis-improving, and sepsis-improving effects
  • Inhibits signaling pathways such as TNF/NF-κB and MAPK
  • Regulates immune homeostasis
  • Can be used in research related to immune, allergic, and inflammatory diseases
Catalog No. 50-004-16168 Supplier Medchemexpress LLC Supplier No. HYN0852100MG
Benzoylpaeoniflorin is an orally active monoterpene glycoside compound that exerts anti-inflammatory, anti-allergic, psoriasis-improving, and sepsis-improving effects by inhibiting signaling pathways such as TNF/NF-κB and MAPK, as well as regulating immune homeostasis. It can be used in research related to immune, allergic, and inflammatory diseases.
  • Orally active monoterpene glycoside compound
  • Exerts anti-inflammatory, anti-allergic, psoriasis-improving, and sepsis-improving effects
  • Inhibits signaling pathways such as TNF/NF-κB and MAPK
  • Regulates immune homeostasis
  • Can be used in research related to immune, allergic, and inflammatory diseases