Learn More
Medchemexpress LLC Benzquinamide hydrochloride | 113-69-9 | 440.96 | 1 MG
SDP
Catalog No. 5000448502
Encompass_Preferred
Benzquinamide hydrochloride is an antiemetic that binds to α2A, α2B, and α2C adrenergic receptors. It also inhibits P-glycoprotein mediated drug efflux, which can potentiate the cytotoxicity of anticancer agents in multidrug resistant cells.
  • Antiemetic properties
  • Binds to α2A, α2B, and α2C adrenergic receptors
  • Inhibits P-glycoprotein mediated drug efflux
  • Potentiates cytotoxicity of anticancer agents in multidrug resistant cells
Catalog No. 50-004-48502 Supplier Medchemexpress LLC Supplier No. HYU00244A1MG
Benzquinamide hydrochloride is an antiemetic that binds to α2A, α2B, and α2C adrenergic receptors. It also inhibits P-glycoprotein mediated drug efflux, which can potentiate the cytotoxicity of anticancer agents in multidrug resistant cells.
  • Antiemetic properties
  • Binds to α2A, α2B, and α2C adrenergic receptors
  • Inhibits P-glycoprotein mediated drug efflux
  • Potentiates cytotoxicity of anticancer agents in multidrug resistant cells