Learn More
Apexbio Technology LLC Benztropine mesylate 132-17-2 1g
SDP
Catalog No. 502240407
Encompass_Preferred
Benztropine mesylate (CAS 132-17-2) is a small molecule inhibitor of the dopamine transporter (DAT) exhibiting an IC50 of 118 nM for DAT inhibition By blocking dopamine reuptake benztropine mesylate modulates synaptic dopamine levels thereby influencing dopaminergic neurotransmission This compound is frequently utilized in biomedical research to investigate the regulation of dopamine signaling study transporter pharmacology and explore mechanisms underlying neuropsychiatric and neurodegenerative disorders that involve dopaminergic dysregulation
This item is not returnable. View return policy
Catalog No. 50-224-0407 Supplier Apexbio Technology LLC Supplier No. B15541000
Benztropine mesylate (CAS 132-17-2) is a small molecule inhibitor of the dopamine transporter (DAT) exhibiting an IC50 of 118 nM for DAT inhibition By blocking dopamine reuptake benztropine mesylate modulates synaptic dopamine levels thereby influencing dopaminergic neurotransmission This compound is frequently utilized in biomedical research to investigate the regulation of dopamine signaling study transporter pharmacology and explore mechanisms underlying neuropsychiatric and neurodegenerative disorders that involve dopaminergic dysregulation