Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benztropine Mesylate, MP Biomedicals™

Catalog No. ICN15362680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15362680 100 mg
ICN15362625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15362680 Supplier MP Biomedicals, Inc Supplier No. 0215362680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132-17-2
Molecular Formula C22H29NO4S
Molecular Weight (g/mol) 403.54
MDL Number MFCD00074784
InChI Key CPFJLLXFNPCTDW-STYNFMPRSA-N
Synonym benztropine mesylate, cogentin, benztropine mesilate, benztropine methanesulfonate, unii-wmj8tl7510, benztropine methylsulfonate, benzatropine mesilate, benzatropine mesylate, benzotropine mesylate, cobrentin methanesulfonate
PubChem CID 3246155
SMILES CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 100 mg
IUPAC Name (1R,5S)-3-benzhydryloxy-8-methyl-8-azabicyclo[3.2.1]octane;methanesulfonic acid
Formula Weight 403.5
Physical Form Solid
Chemical Name or Material Benztropine Mesylate
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.