Learn More
TARGETMOL CHEMICALS INC BENZYDAMINE 50MG
SDP
Catalog No. 502576225
Encompass_Preferred
Also available in 5 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Benzydamine is a benzyl-indazole having analgesic antipyretic and anti-inflammatory effects. Benzydamine is used to reduce post-surgical and post-traumatic pain and edema and to promote healing. purity: 98%
Catalog No. 50-257-6225 Supplier TARGETMOL CHEMICALS INC Supplier No. T880750MG
Also available in 5 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Benzydamine is a benzyl-indazole having analgesic antipyretic and anti-inflammatory effects. Benzydamine is used to reduce post-surgical and post-traumatic pain and edema and to promote healing. purity: 98%