Learn More
Selleck Chemical LLC Benzydamine HCl S4165-50mg
SDP
Supplier:  Selleck Chemical LLC S416550MG
Encompass_Preferred
Benzydamine HCl (AF864) is a topical nonsteroidal anti-inflammatory drug (NSAID) with anti-inflammatory analgesic antipyretic and local anesthetic activity
Catalog No. 50-310-9423