Learn More
Medchemexpress LLC Benzydamine hydrochloride | 132-69-4 | 99.2% | 345.87 | 10 MM 1 ML
SDP
Catalog No. 5000405238
Encompass_Preferred
Benzydamine hydrochloride is an orally administered prostaglandin synthesis inhibitor that possesses anti-inflammatory, analgesic, antipyretic, and antibacterial properties. It can inhibit TNF-α, stabilize cell membranes, and reduce oxidative stress within cells. This specific product is a 10 mM solution in water, ready for reconstitution.
  • Anti-inflammatory, analgesic, antipyretic, and antibacterial properties
  • Inhibits TNF-α and stabilizes cell membranes
  • Reduces oxidative stress within cells
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-004-05238 Supplier Medchemexpress LLC Supplier No. HY30235A10MM/1ML
Benzydamine hydrochloride is an orally administered prostaglandin synthesis inhibitor that possesses anti-inflammatory, analgesic, antipyretic, and antibacterial properties. It can inhibit TNF-α, stabilize cell membranes, and reduce oxidative stress within cells. This specific product is a 10 mM solution in water, ready for reconstitution.
  • Anti-inflammatory, analgesic, antipyretic, and antibacterial properties
  • Inhibits TNF-α and stabilizes cell membranes
  • Reduces oxidative stress within cells
  • Suitable for laboratory centrifugation and storage