Learn More
Cayman Chemical Benzydamine (hydrochloride), Purity: ≥95%, CAS: 132-69-4, 5 g
SDP
Catalog No. 502942180
Encompass_Preferred
An NSAID; decreases the viability of THP-1, Saos-2, and HGF-1 cells (IC50s = 59.08, 30.79, and 52.06 µg/ml, respectively); reduces LPS-induced increases in the levels of IL-6 and TNF-α secreted by THP-1 cells at 10 µg/ml; decreases PGE2 and PGF1α levels in unstimulated or IL-1β- or TNF-α-stimulated primary human gingival fibroblasts at 1 µM; reduces hydrochloric acid levels and increases mucin levels in gastric juices isolated from rats at 100 mg/kg; inhibits carrageenan-induced paw edema in rats at an oral dose of 60 mg/kg or applied topically at 3% (w/w)
Catalog No. 50-294-2180 Supplier Cayman Chemical Supplier No. 398085 G
An NSAID; decreases the viability of THP-1, Saos-2, and HGF-1 cells (IC50s = 59.08, 30.79, and 52.06 µg/ml, respectively); reduces LPS-induced increases in the levels of IL-6 and TNF-α secreted by THP-1 cells at 10 µg/ml; decreases PGE2 and PGF1α levels in unstimulated or IL-1β- or TNF-α-stimulated primary human gingival fibroblasts at 1 µM; reduces hydrochloric acid levels and increases mucin levels in gastric juices isolated from rats at 100 mg/kg; inhibits carrageenan-induced paw edema in rats at an oral dose of 60 mg/kg or applied topically at 3% (w/w)