Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl 2,4-dibromobutyrate

Catalog No. 3406510ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-065-10ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-065-10ML Supplier Sigma Aldrich Supplier No. 3406510ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyl 2,4-dibromobutyrate

Specifications

CAS 50712-74-8
Density 1.63 g/mL (at 20°C (literature))
Molecular Formula C11H12Br2O2
Linear Formula BrCH2CH2CHBrCOOCH2C6H5
Refractive Index n20/D 1.558
MDL Number MFCD01862993
Quantity 100 mg
Synonym 2,4-Dibromobutyric acid benzyl ester
Molecular Weight (g/mol) 336.02
Percent Purity ≥97.0% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.