Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl 2-bromoethyl ether

Catalog No. 4748191G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-481-91G 1 g
47-481-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-481-91G Supplier Sigma Aldrich Supplier No. 4748191G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzyl 2-bromoethyl ether is an organic building block
  • Its enthalpy of vaporization at boiling point (527.15k) has been reported to be 46.499kjoule/mol.building block used to prepare a copper phthalocyanine discotic liquid crystal

Specifications

CAS 1462-37-9
Density 1.36 g/mL (at 25°C (literature))
Boiling Point 254°C (lit.)
Molecular Formula C9H11BrO
Refractive Index n20/D 1.541 (literature)
Linear Formula C6H5CH2OCH2CH2Br
MDL Number MFCD01321307
Quantity 1 g
Molecular Weight (g/mol) 215.09
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.