Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 2-Ethylhexyl Phthalate 95.0+%, TCI America™
SDP

Catalog No. B271425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B271425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B271425G Supplier TCI America Supplier No. B271425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27215-22-1
Molecular Formula C23H28O4
Synonym Benzyl Octyl Phthalate, Phthalic Acid Benzyl 2-Ethylhexyl Ester, Phthalic Acid Benzyl Octyl Ester

Specifications

Color Yellow
Boiling Point 401°C
Quantity 25 g
Formula Weight 368.47
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 2-Ethylhexyl Phthalate
EINECSNumber (3)-1312
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.