Learn More
Medchemexpress LLC Benzyl (2S)-4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-2-(tert-butylcarbamoyl)piperazine-1-carboxylate | 189349-50-6 | MFCD05664727 | 100.0% | 522.60 g/mol | C27H34N6O5 | 10 MG
SDP
Catalog No. 5000211598
Encompass_Preferred
L-765314 is a research compound that selectively antagonizes the α1B adrenergic receptor. It has reported inhibition constants in the low nanomolar range for rat and human receptors and is typically supplied as a solid for laboratory pharmacology and receptor characterization studies.
  • Potent α1B receptor antagonist with low-nanomolar affinity.
  • Selectivity suitable for subtype-specific pharmacology studies.
  • Reported Ki values: 5.4 nM (rat) and 2.0 nM (human).
  • Solid powder form allows flexible storage and formulation.
  • Commonly used for receptor binding, signaling, and cardiovascular research.
Catalog No. 50-002-11598 Supplier Medchemexpress LLC Supplier No. HY10138510MG
L-765314 is a research compound that selectively antagonizes the α1B adrenergic receptor. It has reported inhibition constants in the low nanomolar range for rat and human receptors and is typically supplied as a solid for laboratory pharmacology and receptor characterization studies.
  • Potent α1B receptor antagonist with low-nanomolar affinity.
  • Selectivity suitable for subtype-specific pharmacology studies.
  • Reported Ki values: 5.4 nM (rat) and 2.0 nM (human).
  • Solid powder form allows flexible storage and formulation.
  • Commonly used for receptor binding, signaling, and cardiovascular research.