Learn More
Medchemexpress LLC Benzyl 3-[6-[[2-(butylamino)-1-[3-methoxycarbonyl-4-(2-methoxy-2-oxoethoxy)phenyl] | 831217-40-4 | 98.2% | 931.12 g/mol | C54H66N4O10 | 1MG
SDP
Catalog No. 5000238871
Encompass_Preferred
MAL3-101 is a small-molecule allosteric inhibitor of heat shock protein 70 (HSP70) used in cellular and biochemical research. It inhibits HSP70 ATPase activity by blocking interaction with Hsp40 co-chaperones, perturbing protein folding and proteostasis pathways. The compound is supplied as a high-purity research reagent (CAS 831217-40-4) and is intended for in vitro and preclinical studies only.
  • Allosteric inhibitor of HSP70 ATPase activity.
  • Blocks Hsp40-Hsp70 interaction to disrupt protein folding.
  • Applicable to cellular models studying proteostasis and cancer biology.
  • High reported purity supports reproducible results.
  • Supplied in small quantities suitable for screening and pilot studies.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-38871 Supplier Medchemexpress LLC Supplier No. HY1248051MG
MAL3-101 is a small-molecule allosteric inhibitor of heat shock protein 70 (HSP70) used in cellular and biochemical research. It inhibits HSP70 ATPase activity by blocking interaction with Hsp40 co-chaperones, perturbing protein folding and proteostasis pathways. The compound is supplied as a high-purity research reagent (CAS 831217-40-4) and is intended for in vitro and preclinical studies only.
  • Allosteric inhibitor of HSP70 ATPase activity.
  • Blocks Hsp40-Hsp70 interaction to disrupt protein folding.
  • Applicable to cellular models studying proteostasis and cancer biology.
  • High reported purity supports reproducible results.
  • Supplied in small quantities suitable for screening and pilot studies.
  • For research use only; not for human or veterinary use.