Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 3-Oxopiperazine-1-carboxylate 98.0+%, TCI America™
SDP

Catalog No. B41005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41005G 5 g
B41001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41005G Supplier TCI America Supplier No. B41005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78818-15-2
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.255
MDL Number MFCD00173924
InChI Key BAHFPJFBMJTOPU-UHFFFAOYSA-N
Synonym 4-benzyloxycarbonylpiperazin-2-one, 1-cbz-3-oxopiperazine, 4-benzyloxycarbonyl-2-piperazinone, 4-cbz-piperazinone, 1-z-3-oxopiperazine, 4-cbz-piperazin-2-one, 1-piperazinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxopiperazine-1-carboxylic acid benzyl ester, benzyl 3-oxotetrahydro-1 2h-pyrazinecarboxylate, phenylmethyl 3-oxopiperazinecarboxylate
PubChem CID 736777
IUPAC Name benzyl 3-oxopiperazine-1-carboxylate
SMILES C1CN(CC(=O)N1)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 5 g
Formula Weight 234.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 3-Oxopiperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.