Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl (4-aminobicyclo[2.1.1]hexan-1-yl)carbamate hydrochloride | 1354951-84-0 | MFCD20441712 | 99.6% | 282.77 | C14H19ClN2O2 | 50 MG
SDP

Catalog No. 5000300364
Encompass_Preferred

Benzyl (4-aminobicyclo[2.1.1]hexan-1-yl)carbamate hydrochloride is a small-molecule carbamate hydrochloride used as a drug intermediate and reagent in medicinal chemistry and organic synthesis. It is provided as a solid and characterized for purity and stability to support downstream synthetic applications.

  • Solid form, suitable for storage and handling.
  • CAS number 1354951-84-0 and molecular formula C14H19ClN2O2.
  • Molecular weight 282.77 g/mol.
  • High purity (99.6%) as determined by analytical testing.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); solutions at -80°C (6 months) or -20°C (1 month).
  • Intended as a synthetic intermediate for research use.

Catalog No. 50-003-00364 Supplier Medchemexpress LLC Supplier No. HYW03686050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Benzyl (4-aminobicyclo[2.1.1]hexan-1-yl)carbamate hydrochloride is a small-molecule carbamate hydrochloride used as a drug intermediate and reagent in medicinal chemistry and organic synthesis. It is provided as a solid and characterized for purity and stability to support downstream synthetic applications.

  • Solid form, suitable for storage and handling.
  • CAS number 1354951-84-0 and molecular formula C14H19ClN2O2.
  • Molecular weight 282.77 g/mol.
  • High purity (99.6%) as determined by analytical testing.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); solutions at -80°C (6 months) or -20°C (1 month).
  • Intended as a synthetic intermediate for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.