Learn More
Medchemexpress LLC Benzyl (4-aminobicyclo[2.1.1]hexan-1-yl)carbamate hydrochloride | 1354951-84-0 | MFCD20441712 | 99.6% | 282.77 g/mol | C14H19ClN2O2 | 500 MG
SDP
Catalog No. 5000299709
Encompass_Preferred
Benzyl N-{4-aminobicyclo[2.1.1]hexan-1-yl}carbamate hydrochloride is a carbamate hydrochloride salt used as a chemical intermediate in medicinal chemistry and organic synthesis. It has chemical formula C14H19ClN2O2 and a molecular weight of 282.77 g/mol. The compound is supplied for research use in milligram-to-gram quantities; users should consult the certificate of analysis for storage and handling details.
  • Used as an intermediate in synthesis of active pharmaceutical compounds.
  • Hydrochloride salt form aids solubility and handling.
  • Reported high purity suitable for analytical and synthetic work.
  • Available in milligram and gram pack sizes for research-scale use.
  • Certificate of analysis available for lot-specific purity and storage instructions.
Catalog No. 50-002-99709 Supplier Medchemexpress LLC Supplier No. HYW036860500MG
Benzyl N-{4-aminobicyclo[2.1.1]hexan-1-yl}carbamate hydrochloride is a carbamate hydrochloride salt used as a chemical intermediate in medicinal chemistry and organic synthesis. It has chemical formula C14H19ClN2O2 and a molecular weight of 282.77 g/mol. The compound is supplied for research use in milligram-to-gram quantities; users should consult the certificate of analysis for storage and handling details.
  • Used as an intermediate in synthesis of active pharmaceutical compounds.
  • Hydrochloride salt form aids solubility and handling.
  • Reported high purity suitable for analytical and synthetic work.
  • Available in milligram and gram pack sizes for research-scale use.
  • Certificate of analysis available for lot-specific purity and storage instructions.