Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-Bromophenyl Ketone 97.0+%, TCI America™
SDP

Catalog No. B20205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20205G 5 g
B20201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20205G Supplier TCI America Supplier No. B20205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2001-29-8
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00016331
InChI Key MOSIKPSTRPODHQ-UHFFFAOYSA-N
Synonym benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, acmc-1chtj
PubChem CID 519738
IUPAC Name 1-(4-bromophenyl)-2-phenylethanone
SMILES C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 275.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 4-Bromophenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.