Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-Fluorophenyl Ketone 98.0+%, TCI America™
SDP

Catalog No. B321325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B321325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B321325G Supplier TCI America Supplier No. B321325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 347-84-2
Molecular Formula C14H11FO
Molecular Weight (g/mol) 214.239
MDL Number MFCD00017960
InChI Key YFYKGCQUWKAFLW-UHFFFAOYSA-N
Synonym 4′C-Fluoro-2-phenylacetophenone
PubChem CID 318235
IUPAC Name 1-(4-fluorophenyl)-2-phenylethanone
SMILES C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)F

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 25 g
Formula Weight 214.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 4-Fluorophenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.