Learn More
Medchemexpress LLC Benzyl 4-hydroxybenzoate | 94-18-8 | 99.2% | 228.25 | 25 G
SDP
Catalog No. 5000453250
Encompass_Preferred
Benzyl 4-hydroxybenzoate is a prominent material that can be used as an excipient, such as a bacteriostatic agent or preservative. Pharmaceutical excipients, or pharmaceutical auxiliaries, are inactive ingredients in pharmaceutical preparations.
  • Improves stability, solubility, and processability of pharmaceutical preparations.
  • Affects absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Suitable for research use only.
Catalog No. 50-004-53250 Supplier Medchemexpress LLC Supplier No. 25GHYW013482
Benzyl 4-hydroxybenzoate is a prominent material that can be used as an excipient, such as a bacteriostatic agent or preservative. Pharmaceutical excipients, or pharmaceutical auxiliaries, are inactive ingredients in pharmaceutical preparations.
  • Improves stability, solubility, and processability of pharmaceutical preparations.
  • Affects absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Suitable for research use only.