Learn More
Medchemexpress LLC Benzyl 4-hydroxybenzoate | 94-18-8 | 99.2% | 228.25 | 25 MG
SDP
Catalog No. 5000366397
Encompass_Preferred
Benzyl 4-hydroxybenzoate is a prominent material used as an excipient, specifically as a bacteriostatic agent and preservative. Pharmaceutical excipients are chemical substances, other than pharmaceutical ingredients, used in the pharmaceutical process. They are generally inactive ingredients in pharmaceutical preparations that improve stability, solubility, and processability, and can also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Functions as a bacteriostatic agent
  • Acts as a preservative
  • Improves stability of preparations
  • Enhances solubility of preparations
  • Facilitates processability of preparations
  • Impacts drug absorption, distribution, metabolism, and elimination (ADME)
Catalog No. 50-003-66397 Supplier Medchemexpress LLC Supplier No. 25MGHYW013482S
Benzyl 4-hydroxybenzoate is a prominent material used as an excipient, specifically as a bacteriostatic agent and preservative. Pharmaceutical excipients are chemical substances, other than pharmaceutical ingredients, used in the pharmaceutical process. They are generally inactive ingredients in pharmaceutical preparations that improve stability, solubility, and processability, and can also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Functions as a bacteriostatic agent
  • Acts as a preservative
  • Improves stability of preparations
  • Enhances solubility of preparations
  • Facilitates processability of preparations
  • Impacts drug absorption, distribution, metabolism, and elimination (ADME)