Learn More
Medchemexpress LLC Benzyl 4-hydroxybenzoate | 94-18-8 | 99.2% | 228.25 | 50 G
SDP
Catalog No. 5000454159
Encompass_Preferred
Benzyl 4-hydroxybenzoate is a prominent material and can serve as a pharmaceutical excipient, specifically as a bacteriostatic agent or preservative. Pharmaceutical excipients are generally inactive ingredients in pharmaceutical preparations, designed to enhance the stability, solubility, and processability of these preparations. Beyond these roles, pharmaceutical excipients can also influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Prominent material in pharmaceutical applications.
  • Functions as a bacteriostatic agent.
  • Acts as a preservative.
  • Improves the stability of pharmaceutical preparations.
  • Enhances the solubility of pharmaceutical preparations.
  • Aids in the processability of pharmaceutical preparations.
  • Can affect the ADME processes of co-administered drugs.
Catalog No. 50-004-54159 Supplier Medchemexpress LLC Supplier No. HYW01348250G
Benzyl 4-hydroxybenzoate is a prominent material and can serve as a pharmaceutical excipient, specifically as a bacteriostatic agent or preservative. Pharmaceutical excipients are generally inactive ingredients in pharmaceutical preparations, designed to enhance the stability, solubility, and processability of these preparations. Beyond these roles, pharmaceutical excipients can also influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Prominent material in pharmaceutical applications.
  • Functions as a bacteriostatic agent.
  • Acts as a preservative.
  • Improves the stability of pharmaceutical preparations.
  • Enhances the solubility of pharmaceutical preparations.
  • Aids in the processability of pharmaceutical preparations.
  • Can affect the ADME processes of co-administered drugs.