Learn More
Medchemexpress LLC Benzyl 4-hydroxybenzoate-d4 | 1219805-81-8 | 99.86% | 232.27 | 5 MG
SDP
Catalog No. 5000454230
Encompass_Preferred
Benzyl 4-hydroxybenzoate-d4 is the deuterium labeled Benzyl 4-hydroxybenzoate. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54230 Supplier Medchemexpress LLC Supplier No. HYW013482S5MG
Benzyl 4-hydroxybenzoate-d4 is the deuterium labeled Benzyl 4-hydroxybenzoate. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.