Learn More
Medchemexpress LLC Benzyl 4-hydroxybenzoate-d4 | 1219805-81-8 | 99.86% | 232.27 | 50 MG
SDP
Catalog No. 5000454677
Encompass_Preferred
Benzyl 4-hydroxybenzoate-d4 is the deuterium labeled Benzyl 4-hydroxybenzoate. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54677 Supplier Medchemexpress LLC Supplier No. HYW013482S50MG
Benzyl 4-hydroxybenzoate-d4 is the deuterium labeled Benzyl 4-hydroxybenzoate. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.