Learn More
Medchemexpress LLC Benzyl 4-hydroxybenzoate-d4 | 1219805-81-8 | 99.9% | 232.27 | 10 MG
SDP
Catalog No. 5000454313
Encompass_Preferred
Benzyl 4-hydroxybenzoate-d4 is the deuterium labeled Benzyl 4-hydroxybenzoate. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-54313 Supplier Medchemexpress LLC Supplier No. HYW013482S10MG
Benzyl 4-hydroxybenzoate-d4 is the deuterium labeled Benzyl 4-hydroxybenzoate. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs