Learn More
Medchemexpress LLC Benzyl (5-hydroxypentyl) carbamate | 87905-98-4 | MFCD01863135 | >98.0% | 237.30 g/mol | C13H19NO3 | 1 G
SDP
Catalog No. 5000283844
Encompass_Preferred
Benzyl (5-hydroxypentyl) carbamate is a carbamate-based linker and protecting-group reagent used in organic synthesis and nucleic acid chemistry. It is typically provided as a research-scale solid and is used to introduce a benzyl carbamate moiety or to functionalize hydroxyl-bearing intermediates in medicinal chemistry and DNA/RNA-related syntheses.
  • Used as a carbamate linker or protecting group in synthetic chemistry.
  • Compatible with applications in DNA and RNA synthesis and medicinal chemistry.
  • Contains a terminal hydroxyl group for conjugation or further functionalization.
  • Supplied as a research-scale solid suitable for laboratory use.
Catalog No. 50-002-83844 Supplier Medchemexpress LLC Supplier No. HYW1421941G
Benzyl (5-hydroxypentyl) carbamate is a carbamate-based linker and protecting-group reagent used in organic synthesis and nucleic acid chemistry. It is typically provided as a research-scale solid and is used to introduce a benzyl carbamate moiety or to functionalize hydroxyl-bearing intermediates in medicinal chemistry and DNA/RNA-related syntheses.
  • Used as a carbamate linker or protecting group in synthetic chemistry.
  • Compatible with applications in DNA and RNA synthesis and medicinal chemistry.
  • Contains a terminal hydroxyl group for conjugation or further functionalization.
  • Supplied as a research-scale solid suitable for laboratory use.