Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl acetate

Catalog No. B158051KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B158051KG 1 kg
B15805100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B158051KG Supplier Sigma Aldrich Supplier No. B158051KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyl acetate

Specifications

CAS 140-11-4
Melting Point -51°C (lit.)
Density 1.054 g/mL (at 25°C (literature))
Boiling Point 206°C (lit.)
Molecular Formula C9H10O2
Refractive Index n20/D 1.502 (literature)
Linear Formula CH3COOCH2C6H5
MDL Number MFCD00008712
Quantity 1 kg
Synonym Acetic acid benzyl ester
Molecular Weight (g/mol) 150.17
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.