Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, specified according to requirements of Ph.Eur.

Catalog No. p-4813339
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447000010 1 L Glass bottle
AC447000025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447000010 Supplier Thermo Scientific Chemicals Supplier No. 447000010
Only null left
Add to Cart

CAS: 100-51-6 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15.0°C
Density 1.0440g/mL
Boiling Point 205.0°C
Flash Point 96°C
Packaging Glass bottle
Refractive Index 1.5380 to 1.5410
Linear Formula C6H5CH2OH
Quantity 1 L
Merck Index 15,1127
Specific Gravity 1.044
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride
Viscosity 6.6 mPa.s (20°C)
Formula Weight 108.14
Grade ACS Reagent
Related Substances greater than that of benzylalcohol)
Chemical Name or Material Benzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
EINECSNumber 202-859-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.