Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019996 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9996 5 mL
11-101-8459 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9996 Supplier MilliporeSigma Supelco Supplier No. 084215MLF
Add to Cart
Add to Cart

Benzyl alcohol belongs to the aromatic alcohol group of compounds widely utilized in cosmetics and preservatives. It is also used as a solvent and viscosity-reducing agent.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-51-6
Vapor Pressure 3.75 mmHg (77°C)
Vapor Density 3.7 (vs. air)
Density 1.045 g/mL (at 25°C)
Linear Formula C6H5CH2OH
MDL Number MFCD00004599
Melting Point -16°C to +13°C
Boiling Point 203°C to 205°C
Synonym Benzenemethanol
Quantity 5 mL
Formula Weight 108.14
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.