Learn More
Medchemexpress LLC Benzyl alcohol-d1 (Benzenemethanol-d1) | 14915-25-4 | 95.7% | 100 MG
SDP
Catalog No. 5000408634
Encompass_Preferred
Benzyl alcohol-d1 is a deuterium-labeled benzyl alcohol. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs
  • Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development
Catalog No. 50-004-08634 Supplier Medchemexpress LLC Supplier No. HYB0892S4100MG
Benzyl alcohol-d1 is a deuterium-labeled benzyl alcohol. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs
  • Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development