Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl alcohol-d1 (Benzenemethanol-d1) | 14915-25-4 | 95.7% | 50 MG
SDP

Catalog No. 5000409312
Encompass_Preferred

Benzyl alcohol-d1, also known as Benzenemethanol-d1, is a deuterium-labeled analog of Benzyl alcohol, intended for research use only.

  • Molecular weight of 109.14 and chemical formula of C7H7DO
  • Appears as a colorless to light yellow liquid
  • Incorporates stable heavy isotopes of hydrogen (deuterium)
  • Soluble in DMSO (≥ 175 mg/mL), Ethanol (≥ 100 mg/mL), and water (20 mg/mL with ultrasonication)
  • Serves as a tracer in scientific investigations
  • Employed as an internal standard for quantitative analysis in techniques such as NMR, GC-MS, and LC-MS

Catalog No. 50-004-09312 Supplier Medchemexpress LLC Supplier No. HYB0892S450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl alcohol-d1, also known as Benzenemethanol-d1, is a deuterium-labeled analog of Benzyl alcohol, intended for research use only.

  • Molecular weight of 109.14 and chemical formula of C7H7DO
  • Appears as a colorless to light yellow liquid
  • Incorporates stable heavy isotopes of hydrogen (deuterium)
  • Soluble in DMSO (≥ 175 mg/mL), Ethanol (≥ 100 mg/mL), and water (20 mg/mL with ultrasonication)
  • Serves as a tracer in scientific investigations
  • Employed as an internal standard for quantitative analysis in techniques such as NMR, GC-MS, and LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.