Learn More
Medchemexpress LLC Benzyl alcohol-d5 (Benzenemethanol-d5) | 68661-10-9 | 99.1% | 50 MG
SDP
Catalog No. 5000409366
Encompass_Preferred
Benzyl alcohol-d5 is the deuterium-labeled version of Benzyl alcohol. Benzyl alcohol is an aromatic alcohol, characterized as a colorless liquid with a mild, pleasant aromatic odor. It is intended for research use only and not for patient use.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09366 Supplier Medchemexpress LLC Supplier No. HYB0892S250MG
Benzyl alcohol-d5 is the deuterium-labeled version of Benzyl alcohol. Benzyl alcohol is an aromatic alcohol, characterized as a colorless liquid with a mild, pleasant aromatic odor. It is intended for research use only and not for patient use.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.