Learn More
Medchemexpress LLC Benzyl alcohol-d7 | 71258-23-6 | 99.6% | 100 MG
SDP
Catalog No. 5000409365
Encompass_Preferred
Benzyl alcohol-d7 is the deuterium-labeled Benzyl alcohol, an aromatic alcohol that appears as a colorless liquid with a mild, pleasant aromatic odor. It is intended for research use only and is not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-09365 Supplier Medchemexpress LLC Supplier No. HYB0892S1100MG
Benzyl alcohol-d7 is the deuterium-labeled Benzyl alcohol, an aromatic alcohol that appears as a colorless liquid with a mild, pleasant aromatic odor. It is intended for research use only and is not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs