Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol GMP Multi Compendial, Greenfield

Catalog No. 502946072
Encompass
Change view
Click to view available options
4 x 4 L Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6072 4 x 4 L Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6072 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P303WORLDCS4L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) 108.14

Specifications

Boiling Point 401 °F (205 °C)
Quantity 4x4L Case
Percent Purity 100%
Grade Reagent
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.