Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, MP Biomedicals™

Catalog No. MP219388290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219388290 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219388290 Supplier MP Biomedicals, Inc Supplier No. 0219388290
Add to Cart
Add to Cart

Benzyl alcohol ACS reagent grade, Clear Liquid

  • It may be used for the benzylation of toluene.
  • Benzyl alcohol (BnOH) is a monoaromatic primary alcohol widely used as a solvent and an intermediate in cosmetic formulations, pharmaceutical and flavor/fragrance industries.
  • It shows bacteriostatic property.
  • Commercial method for its synthesis is by the alkaline hydrolysis of benzyl chloride.
  • Conformational studies by ab initio calculations shows gauche cis conformation around the alcoholic group.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym Benzene Carbinol, Benzenemethanol, Benzoyl Alcohol, Hydroxytoluene
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point °C to 15.3°C (literature)
Color ≤20 (APHA)
Density 1.0442g/cm3 at 22.5°C (Lit.)
Boiling Point 205.4°C (at 1013 hPa (Literature))
Flash Point 204.8°F/96°C (Lit.)
Refractive Index 0.03 hPa at 20°C ; 13.3 mmHg ( 100°C), 3.75 mmHg ( 77°C) (Lit.)
Quantity 500 mL
Beilstein 878307
Vapor Density 3.7 (vs air) (Lit.)
Vapor Pressure 0.03 hPa at 20°C ; 13.3 mmHg ( 100°C), 3.75 mmHg ( 77°C) (Lit.)
Grade ACS Reagent
Physical Form Liquid
Assay 0.999
Show More Show Less
RTECSNumber DN3150000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.