Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019546 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9546 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9546 Supplier MilliporeSigma Supelco Supplier No. PHR10191G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Benzyl alcohol is used as a preservative in pharmaceutical preparations in concentration up to 2% and as a solubilizer in concentrations of 5%. It is effective against gram-positive organisms. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-51-6
Ignition Point 817°F
Vapor Pressure 3.75 mmHg (77°C)
Vapor Density 3.7 (vs. air)
Density 1.045 g/mL (at 25°C)
Flash Point 101°C - DIN 51758
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H5 CH2 OH
MDL Number MFCD00004599
Melting Point -16°C to -13°C
Boiling Point 203°C to 205°C
Synonym Benzenemethanol
Type Certified Reference Material
Quantity 1 g
Formula Weight 108.14
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.