Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol, puriss., 99 to 100.5% (GC), Solstice

Catalog No. 60045517 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless steel can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-517 6 x 1 L Glass Bottle
60-045-513 1 L Glass Bottle
60-045-514 2.5 L Glass Bottle
60-045-516 4 x 2.5 L Glass Bottle
60-045-515 30 kg Stainless steel can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-517 Supplier Solstice Supplier No. 241226X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of BP, Ph. Eur., NF, 99-100.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Boiling Range 203° to 205°C (lit.)
Melting Point -16°C to -13°C
Density 1.045g/mL at 25°C
Flash Point 96°C (204.8°F)
Ignition Residue ≤0.005% (as SO4)
Ignition Point 817°F
Packaging Glass Bottle
Linear Formula C6H5CH2OH
Quantity 6 x 1 L
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Vapor Pressure 13.3mmHg (100°C), 3.75mmHg (77°C)
Formula Weight 108.14g/mol
Percent Purity 99 to 100.5% (GC)
Grade puriss.
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, Meets analytical specification of Ph.Eur.BP, NF
Show More Show Less
Hazard Statement P261-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338
Precautionary Statement H302 + H332-H319
EINECSNumber 202-859-9
RTECSNumber DN3150000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.