Learn More
U.S. Pharmacopeia Benzyl Alcohol, 100-51-6, MFCD00004599, 500mg/ampule
Catalog No. NC9187297
Encompass
Molecular Formula C7H8O, Molecular Weight 108.14 g/mol, Melting Point 4.64 °F (-15.2 °C), Boiling Point 401.54 °F (205.3 °C), Synonyms: (Hydroxymethyl)benzene, Phenylmethanol
Catalog No. NC9187297 Supplier U.S. Pharmacopeia Supplier No. 1061901
Molecular Formula C7H8O, Molecular Weight 108.14 g/mol, Melting Point 4.64 °F (-15.2 °C), Boiling Point 401.54 °F (205.3 °C), Synonyms: (Hydroxymethyl)benzene, Phenylmethanol