Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl azide, 94%

Catalog No. AA3263504
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3263504 2 g
AA3263509 10 g
AA3263518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3263504 Supplier Thermo Scientific Chemicals Supplier No. 03263504
Add to Cart

CAS: 622-79-7 | C7H7N3 | 133.154 g/mol

Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-79-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00013836
InChI Key UDLLFLQFQMACJB-UHFFFAOYSA-N
Synonym azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029
PubChem CID 12152
IUPAC Name azidomethylbenzene
SMILES C1=CC=C(C=C1)CN=[N+]=[N-]

Specifications

Density 1.0655
Boiling Point 82°C to 85°C (16 mm)
Flash Point 43°C (110°F)
Assay Percent Range 94%
Refractive Index 1.5341
Quantity 2 g
Solubility Information Insoluble in waterMiscible with ethanol and diethyl ether. Immiscible with water.
Formula Weight 133.15
Physical Form Liquid
Chemical Name or Material Benzyl azide
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
RTECSNumber XS6650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.