Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl Azide Solution

Catalog No. 74243010ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-243-010ML 10 mL
74-243-050ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 74-243-010ML Supplier Sigma Aldrich Supplier No. 74243010ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyl azide is an aromatic azide generally used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.

Specifications

CAS 622-79-7
Molecular Formula C7H7N3
Linear Formula C7H7N3
MDL Number MFCD00013836
Quantity 10 mL
Synonym alpha-Azidotoluene solution; (Azidomethyl)benzene solution
Molecular Weight (g/mol) 133.15 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.