Learn More
Medchemexpress LLC Benzyl benzoate | 120-51-4 | 3075 | 100.0% | 50 G
SDP
Catalog No. 5000407882
Encompass_Preferred
Benzyl benzoate, also known as Phenylmethyl benzoate, is a compound recognized for its diverse biological activities and industrial applications. It functions as an orally active anti-scabies agent, acaricide, and fungicide. Additionally, it acts as an angiotensin II inhibitor, demonstrating antihypertensive effects.
  • Orally active anti-scabies agent
  • Acaricide with an EC50 of 0.06 g/m2
  • Fungicidal properties
  • Inhibits angiotensin II activity
  • Exhibits antihypertensive effects
  • Suitable for use in perfumes
  • Applicable in pharmaceutical formulations
  • Employed in the food industry
Catalog No. 50-004-07882 Supplier Medchemexpress LLC Supplier No. HYB093550G
Benzyl benzoate, also known as Phenylmethyl benzoate, is a compound recognized for its diverse biological activities and industrial applications. It functions as an orally active anti-scabies agent, acaricide, and fungicide. Additionally, it acts as an angiotensin II inhibitor, demonstrating antihypertensive effects.
  • Orally active anti-scabies agent
  • Acaricide with an EC50 of 0.06 g/m2
  • Fungicidal properties
  • Inhibits angiotensin II activity
  • Exhibits antihypertensive effects
  • Suitable for use in perfumes
  • Applicable in pharmaceutical formulations
  • Employed in the food industry