Learn More
Medchemexpress LLC Benzyl benzoate | 120-51-4 | 99.9% | 10 G
SDP
Catalog No. 5000408285
Encompass_Preferred
Benzyl benzoate (Phenylmethyl benzoate) is an orally active anti-scabies agent, acaricide, and fungicide. It acts as an angiotensin II (Ang II) inhibitor, demonstrating antihypertensive effects. This versatile compound finds application in perfumes, pharmaceuticals, and the food industry.
  • Functions as an anti-scabies agent, acaricide, and fungicide
  • Inhibits angiotensin II (Ang II) activity
  • Exhibits antihypertensive effects
  • Utilized in perfumes, pharmaceuticals, and the food industry
  • Causes pathological changes in liver, kidney, thymus, prostate, and epididymal tissues in rats at certain dosages
  • Significantly inhibits Ang II-induced hypertension in mice
Catalog No. 50-004-08285 Supplier Medchemexpress LLC Supplier No. HYB093510G
Benzyl benzoate (Phenylmethyl benzoate) is an orally active anti-scabies agent, acaricide, and fungicide. It acts as an angiotensin II (Ang II) inhibitor, demonstrating antihypertensive effects. This versatile compound finds application in perfumes, pharmaceuticals, and the food industry.
  • Functions as an anti-scabies agent, acaricide, and fungicide
  • Inhibits angiotensin II (Ang II) activity
  • Exhibits antihypertensive effects
  • Utilized in perfumes, pharmaceuticals, and the food industry
  • Causes pathological changes in liver, kidney, thymus, prostate, and epididymal tissues in rats at certain dosages
  • Significantly inhibits Ang II-induced hypertension in mice