Learn More
Medchemexpress LLC Benzyl Benzoate | 120-51-4 | MFCD00003075 | >99.98% | 1 ML
SDP
Catalog No. 5000408948
Encompass_Preferred
Benzyl benzoate is an orally active anti-scabies agent, acaricide, and fungicide. It also acts as an angiotensin II (Ang II) inhibitor with antihypertensive effects. This chemical is suitable for use in perfumes, pharmaceuticals, and the food industry.
  • Orally active anti-scabies agent
  • Functions as an acaricide and fungicide
  • Inhibits angiotensin II (Ang II) activity
  • Exhibits antihypertensive effects
  • Suitable for use in perfumes, pharmaceuticals, and the food industry
Catalog No. 50-004-08948 Supplier Medchemexpress LLC Supplier No. HYB093510MM/1ML
Benzyl benzoate is an orally active anti-scabies agent, acaricide, and fungicide. It also acts as an angiotensin II (Ang II) inhibitor with antihypertensive effects. This chemical is suitable for use in perfumes, pharmaceuticals, and the food industry.
  • Orally active anti-scabies agent
  • Functions as an acaricide and fungicide
  • Inhibits angiotensin II (Ang II) activity
  • Exhibits antihypertensive effects
  • Suitable for use in perfumes, pharmaceuticals, and the food industry