Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyl Benzoate

Catalog No. B66301L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B66301L 1 L
B6630250ML 250 mL
B6630500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B66301L Supplier Sigma Aldrich Supplier No. B66301L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyl Benzoate

Specifications

CAS 120-51-4
Melting Point 17°C to 20°C
Density 1.118 g/mL at 20°C
Boiling Point 323°C to 324°C
Molecular Formula C14H12O2
Linear Formula C6H5COOCH2C6H5
Refractive Index n20/D 1.568
MDL Number MFCD00003075
Quantity 1 L
Synonym Benzoic acid benzyl ester
Molecular Weight (g/mol) 212.24 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.